Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110209.1 | NP_001103679.1 | 3 - 4 | 263 | 107 | |
| NM_027133.3 | NP_081409.1 | 3 - 4 | 263 | 107 | ||
| GenBank mRNA | AK009311.1 | - | 3 - 4 | 237 | 107 | |
| AK014897.1 | - | 3 - 4 | 161 | 107 | ||
| AK053593.1 | - | 3 - 4 | 232 | 107 | ||
| AK082900.1 | - | 3 - 4 | 247 | 107 | ||
| AK136506.1 | - | 3 - 4 | 238 | 107 | ||
| AK137445.1 | - | 3 - 4 | 264 | 107 | ||
| AK143497.1 | - | 3 - 4 | 232 | 107 | ||
| AK173241.1 | - | 3 - 4 | 195 | 107 | ||
| AY312281.1 | - | 3 - 4 | 184 | 107 | ||
| BC057961.1 | - | 3 - 4 | 190 | 5353629 | 107 | |
| BC060153.1 | - | 2 - 3 | 101 | 108 | ||
| BY770278.1 | - | 3 - 4 | 263 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lnp
Genomic Map