Assay Details
| Gene Symbol: Treml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289457.1 | NP_001276386.1 | 5 - 6 | 675 | 107 | |
| NM_027763.2 | NP_082039.1 | 5 - 6 | 690 | 107 | ||
| NR_110338.1 | 5 - 6 | 446 | 107 | |||
| GenBank mRNA | AK017256.1 | - | 5 - 6 | 676 | 107 | |
| AY078502.1 | - | 5 - 6 | 690 | 107 | ||
| AY286071.1 | - | 5 - 6 | 675 | 107 | ||
| BC138399.1 | - | 5 - 6 | 674 | 8861419 | 107 | |
| BC145263.1 | - | 5 - 6 | 674 | 9053816 | 107 | |
| DQ087181.1 | - | 5 - 6 | 443 | 107 | ||
| DQ087182.1 | - | 4 - 5 | 824 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | NM_027763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | AK017256.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | AK079428.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | AY078502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | AY286071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | NM_027763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | AK017256.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | AK079428.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | AY078502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279062 | AY286071.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573623 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102253 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | NM_027763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | AK017256.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | AK079428.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | AY078502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | AY286071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | NM_027763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | AK017256.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | AK079428.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | AY078502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279062 | AY286071.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | NM_027763.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | AK017256.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | AK079428.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | AY078502.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | AY286071.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | NM_027763.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | AK017256.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | AK079428.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | AY078502.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P279062 | AY286071.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31217022 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102253 | - | - |
| Whole Mouse Genome G4122A | A_51_P279062 | NM_027763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | AK017256.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | AK079428.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | AY078502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | AY286071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | NM_027763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | AK017256.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | AK079428.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | AY078502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279062 | AY286071.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102253 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13272064 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13091263 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13272060 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13272063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31216997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31216995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31217030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31216999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13272064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13256741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13224733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13471276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13224737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13416507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13502058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13091260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13484904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13426565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13230074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13190727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13416509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13230069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31217049 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445795 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452630_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445789 | - | - |
| Mouse Genome 430 2.0 Array | 1452630_at | - | - |
| Mouse Genome 430 2.0 Array | 1452630_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71326_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71326_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700009065.mm.1 | NM_001289457.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009065.mm.1 | NM_027763.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009065.mm.1 | NR_110338.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009065.mm.1 | TC1700000938.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700009068.mm.1 | NM_001289451.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009068.mm.1 | NM_001289457.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009068.mm.1 | NM_027763.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009068.mm.1 | NR_110338.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009068.mm.1 | TC1700000938.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700009066.mm.1 | NM_001289451.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009066.mm.1 | NM_001289457.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009066.mm.1 | NM_027763.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009066.mm.1 | NR_110338.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009066.mm.1 | TC1700000938.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700009067.mm.1 | NM_001289451.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009067.mm.1 | NM_001289457.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009067.mm.1 | NM_027763.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009067.mm.1 | NR_110338.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009067.mm.1 | TC1700000938.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700009069.mm.1 | NM_001289451.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009069.mm.1 | NM_001289457.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009069.mm.1 | NM_027763.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009069.mm.1 | NR_110338.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009069.mm.1 | TC1700000938.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452630_at | - | - |
| Mouse Expression Set 430 | 1452630_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452630_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452630_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452630_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452630_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 854091 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2523170 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2523170 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2523169 | - | - |

Treml1
Genomic Map