Assay Details
| Gene Symbol: Spred1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277256.1 | NP_001264185.1 | 4 - 5 | 1078 | 65 | |
| NM_033524.3 | NP_277059.1 | 4 - 5 | 1078 | 65 | ||
| GenBank mRNA | AB063495.1 | - | 4 - 5 | 424 | 65 | |
| AK155065.1 | - | 4 - 5 | 1041 | 65 | ||
| AK155070.1 | - | 4 - 5 | 1080 | 65 | ||
| BC041685.1 | - | 3 - 4 | 392 | 65 | ||
| BC079606.1 | - | 4 - 5 | 699 | 30355455 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spred1
Genomic Map