Assay Details
| Gene Symbol: Fsip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF448787.1 | - | 2 - 3 | 297 | 63 | |
| BC125024.1 | - | 2 - 3 | 189 | 40056533 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fsip1
Genomic Map