Assay Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027225.1 | NP_081501.1 | 11 - 12 | 3530 | 77 | |
| NM_177025.5 | NP_795999.2 | 12 - 13 | 3641 | 77 | ||
| GenBank mRNA | AK037010.1 | - | 13 - 14 | 3635 | 77 | |
| AK052533.1 | - | 11 - 12 | 3479 | 77 | ||
| AK135859.1 | - | 3 - 4 | 1869 | 77 | ||
| AK173076.1 | - | 11 - 12 | 3361 | 77 | ||
| AY308746.1 | - | 12 - 13 | 3641 | 77 | ||
| BC067007.1 | - | 11 - 12 | 3529 | 5702930 | 77 | |
More Information
Set Membership: |
|

Cobll1
Genomic Map