| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200031141.mm.1 |
NM_027225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031141.mm.1 |
NM_027225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031141.mm.1 |
NM_177025.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031141.mm.1 |
NM_177025.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031141.mm.1 |
TC0200003656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200031143.mm.1 |
NM_027225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031143.mm.1 |
NM_027225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031143.mm.1 |
NM_177025.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031143.mm.1 |
NM_177025.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031143.mm.1 |
TC0200003656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200031140.mm.1 |
NM_027225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031140.mm.1 |
NM_027225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031140.mm.1 |
NM_177025.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031140.mm.1 |
NM_177025.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200031140.mm.1 |
TC0200003656.mm.1 |
MTA TC |