Assay Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027225.1 | NP_081501.1 | 4 - 5 | 890 | 127 | |
| NM_177025.5 | NP_795999.2 | 4 - 5 | 890 | 127 | ||
| GenBank mRNA | AK037010.1 | - | 5 - 6 | 881 | 127 | |
| AK052533.1 | - | 4 - 5 | 839 | 127 | ||
| AK144943.1 | - | 5 - 6 | 934 | 127 | ||
| AK164624.1 | - | 5 - 6 | 820 | 127 | ||
| AK173076.1 | - | 4 - 5 | 721 | 127 | ||
| AY308746.1 | - | 4 - 5 | 890 | 127 | ||
| BC067007.1 | - | 4 - 5 | 887 | 5702930 | 127 | |
More Information
Set Membership: |
|

Cobll1
Genomic Map