Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110209.1 | NP_001103679.1 | 12 - 13 | 1231 | 87 | |
| GenBank mRNA | AK014897.1 | - | 12 - 13 | 1128 | 87 | |
| AK137445.1 | - | 12 - 13 | 1231 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lnp
Genomic Map