Assay Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026530.5 | NP_080806.4 | 1 - 2 | 58 | 48 | |
| GenBank mRNA | AK146973.1 | - | 1 - 2 | 43 | 48 | |
| AK160356.1 | - | 1 - 2 | 29 | 48 | ||
| CJ180905.1 | - | 1 - 2 | 60 | 48 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146973.1, AK160356.1, NM_026530.5 transcripts.
Set Membership: |
|

Mpnd
Genomic Map