Assay Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026530.5 | NP_080806.4 | 2 - 3 | 312 | 92 | |
| GenBank mRNA | AK012128.1 | - | 1 - 2 | 267 | 92 | |
| AK146973.1 | - | 2 - 3 | 297 | 92 | ||
| AK160356.1 | - | 2 - 3 | 281 | 92 | ||
| BC017625.1 | - | 1 - 2 | 261 | 92 | ||
| CJ180905.1 | - | 2 - 3 | 314 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpnd
Genomic Map