Assay Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026530.5 | NP_080806.4 | 6 - 7 | 848 | 60 | |
| GenBank mRNA | AK012128.1 | - | 5 - 6 | 803 | 60 | |
| AK146973.1 | - | 6 - 7 | 833 | 60 | ||
| AK160356.1 | - | 6 - 7 | 817 | 60 | ||
| AK172381.1 | - | 5 - 6 | 591 | 60 | ||
| AK185646.1 | - | 1 - 2 | 29 | 60 | ||
| BC017625.1 | - | 5 - 6 | 797 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpnd
Genomic Map