Assay Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026530.5 | NP_080806.4 | 8 - 9 | 998 | 111 | |
| GenBank mRNA | AK012128.1 | - | 7 - 8 | 953 | 111 | |
| AK146973.1 | - | 8 - 9 | 983 | 111 | ||
| AK160356.1 | - | 8 - 9 | 967 | 111 | ||
| AK172381.1 | - | 7 - 8 | 741 | 111 | ||
| AK216749.1 | - | 2 - 3 | 128 | 111 | ||
| BC017625.1 | - | 7 - 8 | 947 | 111 | ||
More Information
Set Membership: |
|

Mpnd
Genomic Map