Assay Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026530.5 | NP_080806.4 | 9 - 10 | 1174 | 92 | |
| GenBank mRNA | AK146973.1 | - | 9 - 10 | 1159 | 92 | |
| AK160356.1 | - | 9 - 10 | 1143 | 92 | ||
| AK172381.1 | - | 8 - 9 | 917 | 92 | ||
| AK185646.1 | - | 4 - 5 | 355 | 92 | ||
| AK216749.1 | - | 3 - 4 | 304 | 92 | ||
| BC017625.1 | - | 8 - 9 | 1123 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpnd
Genomic Map