Assay Details
Gene Symbol: Pot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028370.1 | NP_082646.1 | 6 - 7 | 736 | 103 | |
GenBank mRNA | AK045286.1 | - | 6 - 7 | 687 | 103 | |
AK050727.1 | - | 6 - 7 | 735 | 103 | ||
AK165040.1 | - | 6 - 7 | 713 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
