| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700021480.mm.1 |
NM_028370.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021480.mm.1 |
NM_028370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021480.mm.1 |
TC1700002298.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700021477.mm.1 |
NM_028370.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021477.mm.1 |
NM_028370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021477.mm.1 |
TC1700002298.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700021479.mm.1 |
NM_028370.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021479.mm.1 |
NM_028370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021479.mm.1 |
TC1700002298.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700021481.mm.1 |
NM_028370.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021481.mm.1 |
NM_028370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021481.mm.1 |
TC1700002298.mm.1 |
MTA TC |