Assay Details
Gene Symbol: Pot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028370.1 | NP_082646.1 | 12 - 13 | 1585 | 92 | |
GenBank mRNA | AK045286.1 | - | 12 - 13 | 1595 | 92 | |
AK050727.1 | - | 12 - 13 | 1584 | 92 | ||
AK165040.1 | - | 12 - 13 | 1562 | 92 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
