Assay Details
| Gene Symbol: Vps52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172620.3 | NP_766208.2 | 7 - 8 | 910 | 103 | |
| GenBank mRNA | AK051732.1 | - | 7 - 8 | 981 | 103 | |
| AK052523.1 | - | 7 - 8 | 911 | 103 | ||
| AX775735.1 | - | 7 - 8 | 722 | 103 | ||
| BC063329.1 | - | 6 - 7 | 673 | 30251096 | 103 | |
| KJ650214.1 | - | 7 - 8 | 699 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151248 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151247 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151244 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979927 | AX775735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979927 | AK051732.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979927 | AK052523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979927 | NM_172620.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979927 | AK042520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031217 | AX775735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031217 | BC063329.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031217 | AK190492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031217 | AK051732.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031217 | AK052523.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100295 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979927 | AX775735.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979927 | AK051732.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979927 | AK052523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979927 | NM_172620.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979927 | AK042520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031217 | AX775735.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031217 | BC063329.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031217 | AK190492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031217 | AK051732.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031217 | AK052523.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151258 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08764065 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08764085 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03309720 | - | - |
| Whole Mouse Genome G4122A | A_52_P497085 | AX775735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P497085 | BC063329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P497085 | AK214869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P497085 | AK219244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P497085 | AK051732.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08764085 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13178494 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13178497 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151247 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151248 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13057397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13357011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13404836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13057385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13404837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13178494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13057372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13545467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13178498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13057388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13057380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151259 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444131 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444124 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444128 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444129 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444114 | - | - |
| Mouse Genome 430 2.0 Array | 1437078_at | - | - |
| Mouse Genome 430 2.0 Array | 1437078_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447894_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447894_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224705_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224705_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700005646.mm.1 | NM_172620.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005646.mm.1 | NM_172620.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005646.mm.1 | TC1700000604.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005668.mm.1 | NM_172620.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005668.mm.1 | NM_172620.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005668.mm.1 | TC1700000604.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437078_at | - | - |
| Mouse Expression Set 430 | 1437078_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437078_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437078_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 835249 | - | - |
| ABI Mouse Genome Survey Microarray | 845758 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2486231 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240403 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2486231 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240403 | - | - |

Vps52
Genomic Map