Assay Details
| Gene Symbol: Fam24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018873.1 | - | 1 - 2 | 75 | 73 | |
| AK018966.1 | - | 1 - 2 | 75 | 73 | ||
| BC048642.1 | - | 1 - 2 | 92 | 6772199 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC048642.1 transcripts.
Set Membership: |
|

Fam24a
Genomic Map