Assay Details
| Gene Symbol: Smoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022315.2 | NP_071710.2 | 4 - 5 | 669 | 129 | |
| GenBank mRNA | AJ249901.1 | - | 4 - 5 | 640 | 129 | |
| AK030958.1 | - | 4 - 5 | 638 | 129 | ||
| AK159749.1 | - | 4 - 5 | 636 | 129 | ||
| AK160002.1 | - | 4 - 5 | 636 | 129 | ||
| BC019527.1 | - | 4 - 5 | 616 | 4191849 | 129 | |
More Information
Set Membership: |
|

Smoc2
Genomic Map