Assay Details
| Gene Symbol: Ivd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019826.3 | NP_062800.1 | 7 - 8 | 810 | 88 | |
| GenBank mRNA | AF225989.1 | - | 7 - 8 | 789 | 88 | |
| AK005024.1 | - | 7 - 8 | 829 | 88 | ||
| AK017660.1 | - | 7 - 8 | 796 | 88 | ||
| AK077416.1 | - | 7 - 8 | 818 | 88 | ||
| BC018325.1 | - | 7 - 8 | 800 | 4192286 | 88 | |
| BC027198.1 | - | 7 - 8 | 810 | 4237982 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ivd
Genomic Map