Assay Details
| Gene Symbol: Yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145353.2 | NP_663328.2 | 8 - 9 | 1063 | 75 | |
| GenBank mRNA | AK011822.1 | - | 8 - 9 | 1050 | 75 | |
| AK042644.1 | - | 7 - 8 | 735 | 75 | ||
| AK149957.1 | - | 8 - 9 | 896 | 75 | ||
| BC011161.1 | - | 8 - 9 | 1024 | 75 | ||
| BC019384.1 | - | 8 - 9 | 1044 | 5123736 | 75 | |
| BC100575.1 | - | 7 - 8 | 1131 | 6432108 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | BC100575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | AK149957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | BC011161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | BC019384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | NM_145353.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | BC100575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | AK149957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | BC011161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | BC019384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P487278 | NM_145353.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21787 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322832 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323785 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115554 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | BC100575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | AK149957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | BC011161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | BC019384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | NM_145353.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | BC100575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | AK149957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | BC011161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | BC019384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P487278 | NM_145353.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205962 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115554 | - | - |
| Whole Mouse Genome G4122A | A_52_P423357 | BC100575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423357 | AK149957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423357 | BC011161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423357 | BC019384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P423357 | NM_145353.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | BC100575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | AK149957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | BC011161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | BC019384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | NM_145353.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | BC100575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | AK149957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | BC011161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | BC019384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P487278 | NM_145353.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121038 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132721 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13541357 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13541342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445501 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445500 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445504 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444012_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451284_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445496 | - | - |
| Mouse Genome 430 2.0 Array | 1451284_at | - | - |
| Mouse Genome 430 2.0 Array | 1444012_at | - | - |
| Mouse Genome 430 2.0 Array | 1444012_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451284_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 28064_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 28064_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700008442.mm.1 | NM_145353.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008442.mm.1 | NM_145353.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008442.mm.1 | TC1700000884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700008446.mm.1 | NM_145353.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008446.mm.1 | NM_145353.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008446.mm.1 | TC1700000884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700008443.mm.1 | NM_145353.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008443.mm.1 | NM_145353.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008443.mm.1 | TC1700000884.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451284_at | - | - |
| Mouse Expression Set 430 | 1451284_PM_at | - | - |
| Mouse Expression Set 430 | 1444012_PM_at | - | - |
| Mouse Expression Set 430 | 1444012_at | - | - |
| Murine Genome U74v2 Set | 160211_PM_at | - | - |
| Murine Genome U74v2 Set | 160211_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451284_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451284_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451284_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451284_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444012_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444012_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 856003 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6900341 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5270408 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6900341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900341 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270408 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2649801 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222152 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222152 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2649801 | - | - |
| mouse-6 v1.1 (Illumina) | scl50681.9.1_22-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001656.1_37-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270408 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50681.9.1_22-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001656.1_37-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900341 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270408 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1222152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2649801 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2933228 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2649801 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222152 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2933228 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50681.9.1_22-S | - | - |

Yipf3
Genomic Map