Assay Details
| Gene Symbol: Serpinb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027548.3 | NP_081824.1 | 1 - 2 | 44 | 64 | |
| GenBank mRNA | BC064004.1 | - | 1 - 2 | 37 | 30288309 | 64 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC064004.1, NM_027548.3 transcripts.
Set Membership: |
|

Serpinb7
Genomic Map