Assay Details
| Gene Symbol: Poc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026173.3 | NP_080449.1 | 11 - 12 | 1749 | 109 | |
| GenBank mRNA | AK004769.2 | - | 11 - 12 | 1677 | 109 | |
| AK012926.1 | - | 11 - 12 | 1717 | 109 | ||
| AK098149.1 | - | 11 - 12 | 1642 | 109 | ||
| AK150656.1 | - | 11 - 12 | 1750 | 109 | ||
| BC016527.1 | - | 11 - 12 | 1663 | 3157281 | 109 | |
| BC027118.1 | - | 11 - 12 | 1687 | 5359153 | 109 | |
| BC037631.1 | - | 11 - 12 | 1706 | 4954391 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poc5
Genomic Map