Assay Details
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009483.1 | NP_033509.1 | 17 - 18 | 3037 | 90 | |
| GenBank mRNA | AJ002730.1 | - | 16 - 17 | 2496 | 90 | |
| AK080370.1 | - | 17 - 18 | 3056 | 90 | ||
| AK144654.1 | - | 15 - 16 | 2873 | 90 | ||
| AK162600.1 | - | 1 - 2 | 321 | 90 | ||
| BC027130.1 | - | 5 - 6 | 1484 | 90 | ||
| BC053433.1 | - | 17 - 18 | 3037 | 6516368 | 90 | |
| BC075703.1 | - | 8 - 9 | 1947 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kdm6a
Genomic Map