Assay Details
Gene Symbol: Rad50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009012.2 | NP_033038.2 | 12 - 13 | 2227 | 104 | |
GenBank mRNA | AK037641.1 | - | 11 - 12 | 2070 | 104 | |
AK087982.1 | - | 4 - 5 | 638 | 104 | ||
BC058180.1 | - | 11 - 12 | 2020 | 104 | ||
U66887.1 | - | 12 - 13 | 2205 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
