Assay Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153457.7 | NP_703187.2 | 3 - 4 | 2286 | 74 | |
| GenBank mRNA | AB074899.1 | - | 3 - 4 | 1783 | 74 | |
| AK132364.1 | - | 3 - 4 | 2286 | 74 | ||
| BC030455.1 | - | 3 - 4 | 1838 | 5401661 | 74 | |
| BC053926.1 | - | 3 - 4 | 1914 | 6312477 | 74 | |
| BC058579.1 | - | 3 - 4 | 1948 | 6490092 | 74 | |
| CT010392.1 | - | 3 - 4 | 1783 | 74 | ||
More Information
Set Membership: |
|

Rtn1
Genomic Map