Assay Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007596.2 | NP_001007597.1 | 3 - 4 | 549 | 91 | |
| NM_001286448.1 | NP_001273377.1 | 3 - 4 | 1094 | 91 | ||
| NM_153457.7 | NP_703187.2 | 5 - 6 | 2630 | 91 | ||
| NR_104446.1 | 3 - 4 | 436 | 91 | |||
| GenBank mRNA | AB074899.1 | - | 5 - 6 | 2127 | 91 | |
| AK002997.1 | - | 3 - 4 | 547 | 91 | ||
| AK013731.1 | - | 3 - 4 | 484 | 91 | ||
| AK014358.1 | - | 3 - 4 | 502 | 91 | ||
| AK132364.1 | - | 5 - 6 | 2630 | 91 | ||
| BC027091.1 | - | 3 - 4 | 429 | 91 | ||
| BC030455.1 | - | 5 - 6 | 2182 | 5401661 | 91 | |
| BC053926.1 | - | 5 - 6 | 2258 | 6312477 | 91 | |
| BC058579.1 | - | 5 - 6 | 2292 | 6490092 | 91 | |
| CT010392.1 | - | 5 - 6 | 2127 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC027091.1 transcripts.
Set Membership: |
|

Rtn1
Genomic Map