Assay Details
| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172590.3 | NP_766178.2 | 11 - 12 | 1202 | 80 | |
| GenBank mRNA | AK046853.1 | - | 11 - 12 | 1171 | 80 | |
| AK149993.1 | - | 11 - 12 | 1168 | 80 | ||
| AK194357.1 | - | 2 - 3 | 80 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr41
Genomic Map