Assay Details
| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172590.3 | NP_766178.2 | 1 - 2 | 244 | 80 | |
| GenBank mRNA | AK046853.1 | - | 1 - 2 | 213 | 80 | |
| AK082348.1 | - | 1 - 2 | 245 | 80 | ||
| AK149993.1 | - | 1 - 2 | 210 | 80 | ||
| BB650073.1 | - | 1 - 2 | 240 | 80 | ||
| BC138095.1 | - | 1 - 2 | 150 | 8861114 | 80 | |
| BC145144.1 | - | 1 - 2 | 150 | 9053695 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr41
Genomic Map