Assay Details
| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172590.3 | NP_766178.2 | 7 - 8 | 721 | 78 | |
| GenBank mRNA | AK046853.1 | - | 7 - 8 | 690 | 78 | |
| AK082348.1 | - | 7 - 8 | 722 | 78 | ||
| AK149993.1 | - | 7 - 8 | 687 | 78 | ||
| BC138095.1 | - | 7 - 8 | 627 | 8861114 | 78 | |
| BC145143.1 | - | 7 - 8 | 623 | 78 | ||
| BC145144.1 | - | 7 - 8 | 627 | 9053695 | 78 | |
More Information
Set Membership: |
|

Wdr41
Genomic Map