Assay Details
| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172590.3 | NP_766178.2 | 8 - 9 | 775 | 86 | |
| GenBank mRNA | AK046853.1 | - | 8 - 9 | 744 | 86 | |
| AK082348.1 | - | 8 - 9 | 776 | 86 | ||
| BC138095.1 | - | 8 - 9 | 681 | 8861114 | 86 | |
| BC145143.1 | - | 8 - 9 | 677 | 86 | ||
| BC145144.1 | - | 8 - 9 | 681 | 9053695 | 86 | |
More Information
Set Membership: |
|

Wdr41
Genomic Map