Assay Details
| Gene Symbol: Aldh9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019993.3 | NP_064377.2 | 5 - 6 | 1032 | 74 | |
| GenBank mRNA | AB296386.1 | - | 5 - 6 | 977 | 74 | |
| AF170919.1 | - | 5 - 6 | 777 | 74 | ||
| AK132336.1 | - | 5 - 6 | 817 | 74 | ||
| AK149589.1 | - | 5 - 6 | 818 | 74 | ||
| AK150093.1 | - | 5 - 6 | 775 | 74 | ||
| AK154914.1 | - | 5 - 6 | 1016 | 74 | ||
| AK162354.1 | - | 5 - 6 | 816 | 74 | ||
| AK165730.1 | - | 5 - 6 | 1032 | 74 | ||
| AK168261.1 | - | 5 - 6 | 1010 | 74 | ||
| AK168309.1 | - | 5 - 6 | 1010 | 74 | ||
| AK168878.1 | - | 5 - 6 | 981 | 74 | ||
| BC003297.1 | - | 5 - 6 | 809 | 3583976 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh9a1
Genomic Map