Assay Details
| Gene Symbol: Tex21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159532.1 | NP_001153004.1 | 1 - 2 | 85 | 92 | |
| NM_019784.5 | NP_062758.3 | 1 - 2 | 85 | 92 | ||
| GenBank mRNA | AB000682.1 | - | 1 - 2 | 82 | 92 | |
| AK016448.1 | - | 1 - 2 | 75 | 92 | ||
| AK016471.1 | - | 1 - 2 | 56 | 92 | ||
| BC100432.1 | - | 1 - 2 | 85 | 30917866 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB000682.1, AK016471.1, BC100432.1, NM_001159532.1, NM_019784.5 transcripts.
Set Membership: |
|

Tex21
Genomic Map