Assay Details
| Gene Symbol: Postn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198765.1 | NP_001185694.1 | 2 - 3 | 263 | 83 | |
| NM_001198766.1 | NP_001185695.1 | 2 - 3 | 263 | 83 | ||
| NM_015784.3 | NP_056599.1 | 2 - 3 | 263 | 83 | ||
| GenBank mRNA | AK030756.1 | - | 2 - 3 | 262 | 83 | |
| AK154049.1 | - | 2 - 3 | 261 | 83 | ||
| AY651928.1 | - | 2 - 3 | 233 | 83 | ||
| BC007141.1 | - | 2 - 3 | 256 | 83 | ||
| BC031449.1 | - | 2 - 3 | 232 | 4457222 | 83 | |
| D13664.1 | - | 2 - 3 | 245 | 83 | ||
| E05655.1 | - | 2 - 3 | 259 | 83 | ||
| KM117171.1 | - | 2 - 3 | 227 | 83 | ||
| KM117172.1 | - | 2 - 3 | 227 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Postn
Genomic Map