Assay Details
| Gene Symbol: Sh3bp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133816.2 | NP_598577.1 | 1 - 2 | 136 | 82 | |
| GenBank mRNA | AK028826.1 | - | 1 - 2 | 116 | 82 | |
| AK032634.1 | - | 1 - 2 | 177 | 82 | ||
| AK041504.1 | - | 1 - 2 | 184 | 82 | ||
| BC012284.1 | - | 1 - 2 | 136 | 3978494 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028826.1, AK032634.1, AK041504.1, BC012284.1, NM_133816.2 transcripts.
Set Membership: |
|

Sh3bp4
Genomic Map