Assay Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173392.4 | NP_775568.1 | 14 - 15 | 4143 | 108 | |
| GenBank mRNA | AK080779.1 | - | 10 - 11 | 1952 | 108 | |
| AK122241.1 | - | 14 - 15 | 4124 | 108 | ||
| BC042669.1 | - | 14 - 15 | 4141 | 4235024 | 108 | |
More Information
Set Membership: |
|

Zfyve16
Genomic Map