Assay Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173392.4 | NP_775568.1 | 15 - 16 | 4299 | 77 | |
| GenBank mRNA | AK080779.1 | - | 11 - 12 | 2108 | 77 | |
| AK122241.1 | - | 15 - 16 | 4280 | 77 | ||
| BC042669.1 | - | 15 - 16 | 4297 | 4235024 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfyve16
Genomic Map