Assay Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173392.4 | NP_775568.1 | 7 - 8 | 3218 | 64 | |
| GenBank mRNA | AK080779.1 | - | 3 - 4 | 1074 | 64 | |
| AK122241.1 | - | 7 - 8 | 3199 | 64 | ||
| BC042669.1 | - | 7 - 8 | 3216 | 4235024 | 64 | |
More Information
Set Membership: |
|

Zfyve16
Genomic Map