Assay Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173392.4 | NP_775568.1 | 8 - 9 | 3334 | 151 | |
| GenBank mRNA | AK080779.1 | - | 4 - 5 | 1190 | 151 | |
| AK122241.1 | - | 8 - 9 | 3315 | 151 | ||
| BC042669.1 | - | 8 - 9 | 3332 | 4235024 | 151 | |
More Information
Set Membership: |
|

Zfyve16
Genomic Map