Assay Details
| Gene Symbol: Zcchc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145453.2 | NP_663428.2 | 1 - 2 | 93 | 62 | |
| GenBank mRNA | AK007541.1 | - | 1 - 2 | 40 | 62 | |
| AK044802.1 | - | 1 - 2 | 96 | 62 | ||
| AK051610.1 | - | 1 - 2 | 102 | 62 | ||
| BB656071.1 | - | 1 - 2 | 102 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007541.1, AK044802.1, NM_145453.2 transcripts.
Set Membership: |
|

Zcchc9
Genomic Map