Assay Details
| Gene Symbol: Zcchc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145453.2 | NP_663428.2 | 2 - 3 | 496 | 60 | |
| GenBank mRNA | AK051610.1 | - | 2 - 3 | 505 | 60 | |
| BC010687.1 | - | 1 - 2 | 411 | 60 | ||
| BC024496.1 | - | 1 - 2 | 420 | 4982244 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zcchc9
Genomic Map