Assay Details
| Gene Symbol: Sox30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173384.2 | NP_775560.1 | 2 - 3 | 1303 | 121 | |
| GenBank mRNA | AY005801.1 | - | 2 - 3 | 1303 | 121 | |
More Information
Set Membership: |
|

Sox30
Genomic Map