Assay Details
| Gene Symbol: Ramp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016894.3 | NP_058590.1 | 2 - 3 | 453 | 72 | |
| GenBank mRNA | AB019046.1 | - | 2 - 3 | 264 | 72 | |
| AF146522.1 | - | 2 - 3 | 245 | 72 | ||
| AF209904.1 | - | 2 - 3 | 224 | 72 | ||
| AJ250489.1 | - | 2 - 3 | 245 | 72 | ||
| AK157896.1 | - | 2 - 3 | 243 | 72 | ||
| AK165238.1 | - | 2 - 3 | 229 | 72 | ||
| BC012644.1 | - | 2 - 3 | 210 | 4207524 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ramp1
Genomic Map