Assay Details
| Gene Symbol: Tex35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006825.1 | - | 5 - 6 | 357 | 188 | |
| BC099580.1 | - | 5 - 6 | 352 | 30920417 | 188 | |
| BC115790.1 | - | 5 - 6 | 354 | 40100579 | 188 | |
| BY706810.1 | - | 5 - 6 | 357 | 188 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex35
Genomic Map