Assay Details
Gene Symbol: Tex35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006825.1 | - | 7 - 8 | 600 | 84 | |
BC099580.1 | - | 7 - 8 | 595 | 30920417 | 84 | |
BC115790.1 | - | 7 - 8 | 597 | 40100579 | 84 | |
BY706810.1 | - | 7 - 8 | 600 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953938 | AK006825.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953938 | BC099580.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953938 | NM_028540.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359092 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610542 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828897 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756369 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318400 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798243 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19488 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154824 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153702 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953938 | AK006825.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953938 | BC099580.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953938 | NM_028540.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153702 | - | - |
Whole Mouse Genome G4122A | A_51_P478853 | AK006825.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P478853 | BC099580.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P478853 | BC115790.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P478853 | NM_028540.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153702 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01016497 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00517954 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01065796 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01016501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20746853 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746789 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01097883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746811 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746833 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00171955 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00232744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746793 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00272422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01065795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00299473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00272425 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00575060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746779 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00232742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746801 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746765 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01097884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00171954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00463692 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746762 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00532462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01016500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746832 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00463697 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00846698 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01065800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01097885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746821 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01097881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00575061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00272423 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01016497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00418717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20746744 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10359229 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10359228 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10359226 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10359230 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10359234 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10359227 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10359231 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10359232 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10359233 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1430072_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10359225 | - | - |
Mouse Genome 430 2.0 Array | 1430072_at | - | - |
Mouse Genome 430 2.0 Array | 1430072_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73435_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73435_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100027263.mm.1 | NM_028540.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100027263.mm.1 | TC0100003327.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100027258.mm.1 | NM_028540.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100027258.mm.1 | TC0100003327.mm.1 | MTA TC |
Mouse Expression Set 430 | 1430072_at | - | - |
Mouse Expression Set 430 | 1430072_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430072_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430072_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6370348 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247752 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1247752 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1247752 | - | - |
