Assay Details
| Gene Symbol: Hypm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026096.1 | NP_080372.1 | 1 | 460 | 75 | |
| GenBank mRNA | AK006319.1 | - | 1 | 449 | 75 | |
| AK006788.1 | - | 1 | 460 | 75 | ||
| BC049569.1 | - | 1 | 418 | 6772969 | 75 | |
| BC059721.1 | - | 1 | 435 | 6742451 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | NM_026096.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | AK006319.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | AK006788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | BC049569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | BC059721.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | NM_026096.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | AK006319.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | AK006788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | BC049569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322814 | BC059721.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159600 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | NM_026096.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | AK006319.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | AK006788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | BC049569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | BC059721.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | NM_026096.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | AK006319.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | AK006788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | BC049569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322814 | BC059721.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32248491 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159600 | - | - |
| Whole Mouse Genome G4122A | A_51_P322814 | NM_026096.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | AK006319.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | AK006788.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | BC049569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | BC059721.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | NM_026096.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | AK006319.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | AK006788.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | BC049569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322814 | BC059721.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130711 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14949970 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14949980 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14949987 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14949966 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14949967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14949973 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14949990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32248512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15067301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15067314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14949993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15041605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14949990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15067304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14556218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14949970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15094470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32248505 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10598600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598599 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443743_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420424_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598598 | - | - |
| Mouse Genome 430 2.0 Array | 1420424_at | - | - |
| Mouse Genome 430 2.0 Array | 1443743_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420424_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443743_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67334_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67334_at | - | - |
| Mouse Expression Set 430 | 1420424_PM_at | - | - |
| Mouse Expression Set 430 | 1420424_at | - | - |
| Mouse Expression Set 430 | 1443743_PM_at | - | - |
| Mouse Expression Set 430 | 1443743_at | - | - |
| Murine Genome U74v2 Set | 167606_PM_r_at | - | - |
| Murine Genome U74v2 Set | 167606_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420424_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420424_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420424_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420424_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443743_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443743_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 500314 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2060369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240442 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240442 | - | - |
| mouse-6 v1.1 (Illumina) | scl55049.1.43_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl55049.1.43_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240442 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240442 | - | - |

Hypm
Genomic Map