Assay Details
Gene Symbol: Grhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161406.1 | NP_001154878.1 | 3 - 4 | 430 | 87 | |
GenBank mRNA | AF411212.1 | - | 3 - 4 | 401 | 87 | |
AK076307.1 | - | 3 - 4 | 415 | 87 | ||
AK161048.1 | - | 3 - 4 | 312 | 87 | ||
AK167035.1 | - | 3 - 4 | 430 | 87 | ||
BC013080.1 | - | 3 - 4 | 370 | 3499860 | 87 | |
BC037233.1 | - | 3 - 4 | 422 | 4951512 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
