Assay Details
| Gene Symbol: Usp9x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009481.2 | NP_033507.2 | 2 - 3 | 488 | 98 | |
| GenBank mRNA | AK028443.1 | - | 2 - 3 | 473 | 98 | |
| AK049283.1 | - | 3 - 4 | 601 | 98 | ||
| DQ086491.1 | - | 2 - 3 | 449 | 98 | ||
| U67874.1 | - | 2 - 3 | 449 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp9x
Genomic Map