Assay Details
| Gene Symbol: Zp3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009581.2 | NP_033607.2 | 2 - 3 | 179 | 77 | |
| GenBank mRNA | BB615894.1 | - | 2 - 3 | 179 | 77 | |
| BC119121.1 | - | 2 - 3 | 169 | 8733870 | 77 | |
| BC119123.1 | - | 2 - 3 | 169 | 8733872 | 77 | |
| U17108.1 | - | 2 - 3 | 177 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zp3r
Genomic Map