Assay Details
Gene Symbol: Zp3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009581.2 | NP_033607.2 | 7 - 8 | 920 | 82 | |
GenBank mRNA | BC119121.1 | - | 7 - 8 | 910 | 8733870 | 82 |
BC119123.1 | - | 7 - 8 | 910 | 8733872 | 82 | |
U17108.1 | - | 7 - 8 | 918 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
